missing translation for 'onlineSavingsMsg'
Learn More

PIGF Antibody (RM0010-8F09), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NB110-85468AF750

Product Code. 18231844

  • £462.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PIGF Monoclonal specifically detects PIGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PIGF
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
PIGF
Mouse recombinant PlGF protein.
0.1 mL
Primary
This recognizes mouse PlGF.
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
RM0010-8F09
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
GPI11 homolog, MGC32646, MGC33136, phosphatidylinositol glycan anchor biosynthesis, class F, phosphatidylinositol glycan, class F, phosphatidylinositol-glycan biosynthesis class F protein, PIG-F
Rat
Protein G purified
Biologically Active Proteins
5281.0
Mouse
IgG2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.