missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIGC Monoclonal antibody specifically detects PIGC in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | PIGC |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1G2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_002633 |
| Gene Alias | EC 2.4.1.198, GPI2class C protein, MGC2049, phosphatidylinositol glycan anchor biosynthesis, class C, phosphatidylinositol glycan, class C |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?