missing translation for 'onlineSavingsMsg'
Learn More

PIGB Antibody (2G7), Novus Biologicals™

Product Code. 18397068 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397068 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397068 Supplier Novus Biologicals Supplier No. H00009488M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIGB Monoclonal antibody specifically detects PIGB in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGB
Applications ELISA
Classification Monoclonal
Clone 2G7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004846
Gene Alias EC 2.4.1, EC 2.4.1.-, GPI mannosyltransferase 3, GPI mannosyltransferase III, GPI-MT-III, MGC21236, phosphatidylinositol glycan anchor biosynthesis, class B, phosphatidylinositol glycan, class B, Phosphatidylinositol-glycan biosynthesis class B protein, PIG-B
Host Species Mouse
Immunogen PIGB (NP_004846, 80 a.a. ~ 172 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TSFVPDEYWQSLEVSHHMVFNYGYLTWEWTERLRSYTYPLIFASIYKILHLLGKDSVQLLIWIPRLAQALLSAVADVRLYSLMKQLENQEVAR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9488
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.