missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527740
This item is not returnable.
View return policy
Description
Recommended positive controls: NIH-3T3. Predicted reactivity: Mouse (82%), Rat (83%), Chicken (85%), Rhesus Monkey (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein required for synthesis of N-acetylglucosaminyl phosphatidylinositol (GlcNAc-PI), the first intermediate in the biosynthetic pathway of GPI anchor. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. Paroxysmal nocturnal hemoglobinuria, an acquired hematologic disorder, has been shown to result from mutations in this gene. Alternate splice variants have been characterized.
Specifications
| PIGA | |
| Polyclonal | |
| Unconjugated | |
| PIGA | |
| AI194334; class A GlcNAc-inositol phospholipid assembly protein; GLCNAC-PI synthesis protein; GPI anchor biosynthesis; GPI3; MCAHS2; N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein; phosphatidylinositol glycan anchor biosynthesis class A; phosphatidylinositol glycan anchor biosynthesis, class A; phosphatidylinositol glycan anchor biosynthesis, class A (paroxysmal nocturnal hemoglobinuria); phosphatidylinositol glycan, class A; phosphatidylinositol N-acetylglucosaminyltransferase subunit A; Phosphatidylinositol-glycan biosynthesis class A protein; phosphatidylinositol-glycan biosynthesis, class A protein; phosphatidylinositolglycan class A; PIGA; PIG-A; PNH1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18700, 5277 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| 0.1M tris glycine with 1% BSA, 10% glycerol and 0.01% thimerosal; pH 7 | |
| P37287, Q64323 | |
| PIGA | |
| Recombinant protein encompassing a sequence within the center region of human PIGA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction