missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIG3 Antibody (OTI3B11), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73400APC
This item is not returnable.
View return policy
Description
PIG3 Monoclonal antibody specifically detects PIG3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PIG3 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI3B11 | |
| Western Blot, Immunohistochemistry | |
| p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3 | |
| Full length human recombinant protein of human TP53I3(NP_671713) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis | |
| 9540 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction