missing translation for 'onlineSavingsMsg'
Learn More

Pifithrin-μ, MedChemExpress

Product Code. 16444298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444298 Supplier Medchem Express Supplier No. HY10940
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pifithrin-μ is an inhibitor of p53 and HSP70, with antitumor and neuroprotective activity.

Pifithrin-μ is an inhibitor of p53 and HSP70, with antitumor and neuroprotective activity.

Specifications

Chemical Name or Material Pifithrin-μ
CAS 64984-31-2
Color White
Purity Grade Notes Research
Molecular Formula C8H7NO2S
Quantity 10 mg
Synonym PFTμ 2-Phenylethynesulfonamide
Solubility Information DMSO : ≥ 108 mg/mL (595.99 mM)
SMILES O=S(C#CC1=CC=CC=C1)(N)=O
Molecular Weight (g/mol) 181.21
Formula Weight 181.21
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.