missing translation for 'onlineSavingsMsg'
Learn More

Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals

Product Code. 15436299
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436299 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436299 Supplier Thermo Scientific Alfa Aesar Supplier No. J64277.#0
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

A cell-permeable p53 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 389850-21-9
Color Brown
Molecular Formula C15H16BrN3O3S
MDL Number MFCD00398180
Quantity 5 mg
Sensitivity Hygroscopic; Light sensitive
Synonym pifithrin-alpha, p-nitro, 1-4-nitrophenyl-2-4,5,6,7-tetrahydro-2-imino-3 2h-benzothiazolyl ethanone, hbr, pifithrin-alpha, p-nitro hydrobromide, 2-2-imino-4,5,6,7-tetrahydro-1,3-benzothiazol-3-yl-1-4-nitrophenyl ethanone hydrobromide, 2-2-imino-4,5,6,7-tetrahydrobenzo d thiazol-3 2h-yl-1-4-nitrophenyl ethanone hbr, 2-2-imino-4,5,6,7-tetrahydrobenzo d thiazol-3 2h-yl-1-4-nitrophenyl ethanone hydrobromide
Solubility Information Soluble in DMSO or ethanol
InChI Key NXPCMOGORSWOLH-UHFFFAOYSA-N
SMILES C1CCC2=C(C1)N(C(=N)S2)CC(=O)C3=CC=C(C=C3)[N+](=O)[O-].Br
IUPAC Name 2-(2-imino-4,5,6,7-tetrahydro-1,3-benzothiazol-3-yl)-1-(4-nitrophenyl)ethanone;hydrobromide
Molecular Weight (g/mol) 398.275
PubChem CID 11553038
Formula Weight 398.3
Physical Form Solid
Chemical Name or Material Pifithrin-alpha, p-Nitro hydrobromide
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.