missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIF1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97931B
This item is not returnable.
View return policy
Description
PIF1 Polyclonal specifically detects PIF1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).
Specifications
| PIF1 | |
| Polyclonal | |
| Immunohistochemistry 1:300, Immunohistochemistry-Paraffin 1:300, Western Blot (Negative), ICC/IF (Negative) | |
| Q9H611 | |
| PIF1 | |
| A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) [UniProt Q9H611] | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 80119 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| C15orf20, PIF, PIF1 5-to-3 Prime DNA helicase homolog, RRM3 DNA helicase-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction