missing translation for 'onlineSavingsMsg'
Learn More

PIEZO2 Antibody, Novus Biologicals™

Product Code. 18421802 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421802 0.1 mL 0.1mL
18425322 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18421802 Supplier Novus Biologicals Supplier No. NBP189893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIEZO2 Polyclonal antibody specifically detects PIEZO2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIEZO2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ESEKPIFTMSAQQSQLKVMDQQSFNKFIQAFSRDTGAMQFLENYEKEDITVAELEGNSNSLWTISPPSKQKMIHELLDPNSSFSVVFSWSIQRNLSLGAKSEIATDKLSFPLKNITRKNIAKM
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 63895
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.