missing translation for 'onlineSavingsMsg'
Learn More

PIEZO2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18426682
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426682 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426682 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP178624MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIEZO2 Polyclonal antibody specifically detects PIEZO2 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIEZO2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1600-1650) [UniProt# Q9H5I5]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 63895
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.