missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIEZO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116998
This item is not returnable.
View return policy
Description
Positive Samples: U-87MG, LO2, MCF7.
Piezo1 is a pore-forming subunit of a mechanosensitive non-specific cation channel, and generates currents characterized by a linear current-voltage relationship that are sensitive to ruthenium red and gadolinium. Structurally, Piezo1 contains 36 transmembrane domains and functions as a homotetramer. Piezo1 plays a key role in epithelial cell adhesion by maintaining integrin activation through R-Ras recruitment to the ER, most probably in its activated state, and subsequent stimulation of calpain signaling. In the kidney, Piezo1 may contribute to the detection of intraluminal pressure changes and to urine flow sensing. Piezo1 acts as shear-stress sensor that promotes endothelial cell organization and alignment in the direction of blood flow through calpain activation and plays a key role in blood vessel formation and vascular structure in both development and adult physiology. The protein encoded by the Piezo1 gene is a mechanically-activated ion channel that links mechanical forces to biological signals. Defects in this gene have been associated with dehydrated hereditary stomatocytosis. Diseases associated with PIEZO1 include Dehydrated Hereditary Stomatocytosis with/without Pseudohyperkalemia And/Or Perinatal Edema and Lymphedema.
Specifications
| PIEZO1 | |
| Polyclonal | |
| Unconjugated | |
| Piezo1 | |
| 9630020g22; DHS; Fam38a; family with sequence similarity 38, member A; KIAA0233; LMPH3; Membrane protein induced by beta-amyloid treatment; Mib; mKIAA0233; piezo type mechanosensitive ion channel component 1; Piezo1; piezo-type mechanosensitive ion channel component 1; Protein FAM38A; protein PIEZO1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 234839, 361430, 9780 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.3 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| E2JF22, Q0KL00, Q92508 | |
| Piezo1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction