missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Zinc Reversible Stain Kit

Product Code. 10168213 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 L
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10168213 1.5 L 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10168213 Supplier Thermo Scientific™ Supplier No. 24582

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Visualize proteins in polyacrylamide gels immediately after electrophoresis and then erase the stain for subsequent transfer or band extraction.

Thermo Scientific™ Pierce Zinc Reversible Stain Kit provides rapid and highly sensitive detection of protein in polyacrylamide gels immediately after electrophoresis as well as erasability for subsequent transfer or band extraction.

This zinc-based protein gel stain provides rapid detection of proteins after their separation by SDS-PAGE. The method is as sensitive as most silver staining techniques in that it detects protein at less than 1ng per band, yet requires less than 15 minutes to complete. The simple, two-step staining protocol results in a stained gel with an opaque milky-white background and clear protein bands. The clear bands of protein are visible when the gel is placed against a dark background.

The Pierce Zinc Reversible Stain Kit is an ideal option for in-gel protein detection preceding many downstream applications, because the zinc-stain is completely reversible. The white background is easily removed and the gel is restored to its original condition with the kit-supplied eraser solution or a Tris-glycine buffer. Afterwards, the proteins are free of any dyes or other contaminants and can be restained by another method, transferred to membrane for Western blotting or eluted from an excised gel piece.

Highlights:

  • Sensitive – detect 0.25ng of protein per band in a mini gel

  • Fast – the entire protocol is complete in 15 minutes

  • Easy – ready-to-use stain and developer solutions are supplied, and no fixing or washing steps are required

  • Gentle – no chemical modification of the proteins occurs; protein bands are visible because the background is stained, not the bands themselves

  • Reversible – completely erase the stain from the gel to easily recover proteins or proceed with other methods

Includes:

Zinc stain, developer, and eraser solutions (500mL each)

Recommended for:

Staining preceding protein recovery for antibody generation or immunological detection; Staining before excising gel pieces for protein digestion and analysis by mass spectrometry; Staining before electrophoretic transfer to membrane for Western blotting; Quick purity checks before proceeding with purification or transfer for Western blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 20 mini gels (SDS-PAGE)
• Zinc Stain, 500 mL
• Zinc Developer, 500 mL
• Zinc Eraser, 500 mL

Store kit at room temperature.
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye Proprietary Mix
Quantity 1.5 L
Target Molecule Protein
Product Line Pierce
Product Type Reversible Protein Gel Stain Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.