Learn More
Thermo Scientific™ Pierce™ TiO2 Magnetic Phosphopeptide Enrichment Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Titanium Dioxide Magnetic Phosphopeptide Enrichment Kit is for isolating phosphopeptides from complex biological samples using titanium dioxide-coated magnetic beads.
The high-performance iron oxide superparamagnetic particles are validated and optimized for use with high-throughput magnetic platforms, such as the Thermo Scientific KingFisher™ 96 and KingFisher Flex Instruments. The beads also enable premium performance for simple benchtop applications using an appropriate magnetic stand.
Highlights:
Complete MS-compatible kits – include ready-to-use binding, wash and elution buffers that are optimized for phosphopeptide enrichment and downstream analysis by MALDI and ESI mass spectrometry
- Optimized for HTS – procedure validated for processing 1 to 96 samples at a time; complete entire assay in about 15 minutes using a KingFisher Flex Instrument
Stable affinity ligand – titanium dioxide is specially coated as a film on the magnetic particles
Selective – affinity system is selective for phosphorylated Ser, Tyr and Thr; exhibits minimal non-specific binding to acidic residues
Sensitive – affinity provides more than 1000 times greater sensitivity than traditional IMAC technologies; enables enrichment and MS-measurement of less than 100 fmol of phosphoprotein
Includes:
Pierce TiO2 Magnetic Beads; Binding, wash and elution buffers; 96-well PCR plates for automated processing
Requires:
Thermo Scientific KingFisher 96 or KingFisher Flex Instrument or other magnetic separator device
Specifications
Specifications
| Description | Pierce Titanium Dioxide Magnetic Phosphopeptide Enrichment Kit, Trial Size |
| Formulation | Multi-component kit with TiO2 magnetic beads |
| Kit Contents | Washing Buffer, 25mL, Thermo-Fast 96 Robotic PCR Plates, 0.2mL wells, 2 plates, Binding Buffer, 100mL, Elution Buffer, 3mL, TiO2 Magnetic Beads (20X), 0.25mL |
| Support Type | Magnetic Bead |
| Quantity | 24 Reaction Kit |
| Sufficient For | Purifying 24 x 100ug peptide samples |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.