missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Silver Stain Rescue Reagent

Product Code. 11861375 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
40 mL
Unit Size:
40mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11861375 40 mL 40mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11861375 Supplier Thermo Scientific™ Supplier No. 24614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Remove undesirable background from unevenly and overly developed silver-stained polyacrylamide electrophoresis gels.

Thermo Scientific™ Pierce Silver Stain Rescue Reagent enables removal of undesirable background from unevenly and overly developed silver-stained polyacrylamide electrophoresis gels.

This simple, two-reagent kit recovers and corrects overdeveloped silver-stained gels by uniformly removing developed silver. By slowly reversing the development process over several minutes, the Pierce Silver Stain Rescue Reagent allows one to monitor and stop destaining when the level is best for the bands of interest. In most silver staining procedures for protein gels, addition of stop-solution must be carefully timed to achieve optimal staining results; frequently, otherwise useful SDS-PAGE data are obscured by over-development at this critical step. The Silver Stain Rescue Reagent can recover over-stained gels, eliminating the need to repeat the experiment (run and stain a new gel). Most excessively stained mini gels can be corrected in less than one hour, depending on the working concentration selected.

Highlights:

  • Easy-to-use – mix the reagents at the recommended dilution and it is ready to rescue silver-stained gels that have dark backgrounds as a result of over-development

  • Simple and effective – eliminates background signal in minutes, allowing speedy but controlled reversal of the staining process to meet your objective; complete the entire process in approx. one hour

  • Saves time and money – procedure is much simpler, many times faster, and significantly less costly than repeating gel-electrophoresis and staining

  • Preserves valuable data – uniformly removes silver from the entire polyacrylamide gel, enabling improvement of band visibility without altering or manipulating the data

  • Compatible with any silver stain – removes high background or non-uniform staining, regardless of the silver stain formulation, whether commercial or homemade

Includes:

Reagent solutions A and B (20mL each)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Kit Contents:
• Silver Stain Rescue Reagent A, 20 ml
• Silver Stain Rescue Reagent B, 20 ml

Storage: Upon receipt store at room temperature.
Detection Location In-Gel Detection
Detection Method Colorimetric
Quantity 40 mL
Target Molecule Protein
Product Line Pierce
Product Type Stain Rescue Reagent

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.