Learn More
Description
Thermo Scientific™ SILAC Protein Quantitation Kits are used for the specific analysis of protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC).
Highlights:
Efficient – 100% label incorporation into proteins of living cells
Reproducible – eliminates intra-experimental variability caused by differential sample preparation
Flexible – media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through dual amino acid isotope labeling
Compatible – label proteins expressed in a wide variety of mammalian cell lines adapted to grow in DMEM, RPMI-1640 or DMEM:F12 medium, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, Jurkat and others
Optimized, specialized media – the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry
High-quality supplements – heavy amino acids with >99% isotope purity; the Stem Cell Screened Dialyzed FBS and Serum Substitute for Mouse Embryonic Stem Cells are tested to ensure that they do not promote stem cell differentiation
Recommended for:
Quantitative analysis of relative changes in protein abundance from different cell treatments; Quantitative analysis of proteins for which there are no antibodies available; Protein expression profiling of normal vs. disease cells; Identification and quantification of hundreds to thousands of proteins in a single experiment
Specifications
Specifications
| Formulation | Multi-component kit with heavy and light L-Lysine and DMEM:F12 Media |
| Kit Contents | L-Lysine-2HCl, 50mg, Dialyzed FBS, 2 x 50mL, DMEM:F12 Media for SILAC, 2 x 500mL, L-Arginine-HCl, 2 x 50mg, L-Lysine-2HCl, 13C6, 50mg |
| Product Type | Protein Quantitation Kit - DMEM:F12 |
| Sufficient For | One complete SILAC experiment (stable isotope labeling with amino acids in cell culture) |
| Quantity | 1 Use Kit |
| For Use With (Application) | SILAC experiments (stable isotope labeling with amino acids in cell culture) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.