missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SILAC Media and Sera

Product Code. 10404205 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
Product Type:
DMEM
DMEM:F12
Dialyzed FBS
RPMI
Unit Size:
500mL
50mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
10404205 500 mL DMEM:F12 500mL
12116490 500 mL Dialyzed FBS 500mL
10423425 500 mL RPMI 500mL
10107883 500 mL DMEM 500mL
10511404 50 mL Dialyzed FBS 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10404205 Supplier Thermo Scientific™ Supplier No. 88215

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).

These Thermo Scientific Pierce cell culture media and serum components are optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of heavy 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Highlights:

  • Flexible–liquid media deficient in both L-lysine and L-arginine and powdered media deficient in L-leucine, L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling

  • Compatible–DMEM, MEM, RPMI-1640, IMDM, Ham's F12 and McCoy's 5A media and dialyzed FBS are used to culture a variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, K562 and Jurkat

  • Optimized, specialized media–the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry

  • High quality–Stem Cell Screened Dialyzed FBS is tested to ensure that it does not promote stem cell differentiation

TRUSTED_SUSTAINABILITY

Specifications

Description DMEM:F12 (1:1) Media for SILAC
Formulation DMEM:F12 (1:1) media minus L-Lysine and L-Arginine for induced pluripotent cells
Product Type DMEM:F12
Sufficient For Part of 1 SILAC experiment
Quantity 500 mL
For Use With (Application) SILAC experiments (stable isotope labeling with amino acids in cell culture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.