Learn More
Invitrogen™ S6 Multispecies In-Cell ELISA Kit, Near Infrared
Description
The S6 Multispecies In-Cell ELISA Near Infrared Detection Kits provide a simple and convenient method for quantifying intracellular proteins in whole cells. Principle of the method This kit enables simultaneous detection of post-translational-modified protein (PTM) and the corresponding unmodified protein, or two different targets within the same well. Simultaneously detecting targets within an ELISA well eliminates variability caused by differences in cell plating. The expression levels of the protein(s) are monitored using primary antibodies specific to the targets and corresponding species-specific near-infrared Thermo Scientific DyLight-Conjugated Secondary Antibodies. Relative protein activation or inactivation is determined as a ratio of modified protein levels to the corresponding unmodified protein measured using a modification-specific antibody and an antibody to unmodified protein, respectively. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P62753 |
| Description | Pierce S6 Near Infrared In-Cell ELISA Kit |
| Conjugate | Unconjugated |
| Immunoassay Kit Format | Multi-component kit with fluorescent conjugates |
| Product Type | ELISA Kit |
| Sample Type | Supernatant |
| Sufficient For | 96 microplate wells of overnight-cultured cells |
| Target Species | Human |
| For Use With (Equipment) | Fluorescent Microplate Reader |
| Gene ID (Entrez) | 6194 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.