missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ S6 Multispecies In-Cell ELISA Kit, Colorimetric

Product Code. 11823855
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11823855

Brand: Invitrogen™ 62207

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In-cell ELISA Kit

The S6 Multispecies In-Cell ELISA Colorimetric Detection Kit is a simple method for quantifying intracellular proteins in whole cells. Principle of the method To perform the assay, cells are first plated, treated and fixed. Expression of the protein(s) of interest is monitored in wells of a microplate using target-specific primary antibodies (see the Important Product Information section for antibodies included in each kit) and a horseradish peroxidase (HRP)-conjugated detection reagent. The kit is supplied with a whole-cell stain to control for differences in cell plating, which is important when measuring relative levels of a protein with different treatments or assessing its post-translational modification (PTM) form. After staining, the results are analyzed by normalizing the absorbance (HRP activity) values to cell number, which adjusts for the cell plating differences among the wells. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P62753
Description Pierce S6 Colorimetric In-Cell ELISA Kit
Conjugate HRP
Immunoassay Kit Format Multi-component kit with HRP-conjugate
Product Type ELISA Kit
Sample Type Supernatant
Sufficient For 96 microplate wells of overnight-cultured cells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6194
Gene Symbol S6
Kit Contents Blocking Buffer, 20X Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, HRP Conjugate, TMB Substrate, TMB Stop Solution, Janus Green Whole-Cell Stain, Elution Buffer, Thin Plate Seal Assembly, Components included only in the target-specific kits:Antibody #1 and Antibody #2, Anti-S6 Antibody
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Supernatant, 10,000 cells
Storage Requirements 2°C to 8°C
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 24 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.