missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ RNA 3' End Desthiobiotinylation Kit

Product Code. 13434209 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13434209 20 Reactions 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13434209 Supplier Thermo Scientific™ Supplier No. 20163
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform easy and efficient desthiobiotin labeling of RNA for use as probes or as antibody alternatives for enrichment of RNA binding proteins.

Perform easy and efficient desthiobiotin labeling of RNA for use as probes or as antibody alternatives for enrichment of RNA binding proteins with Thermo Scientific™ Pierce RNA 3' End Desthiobiotinylation Kit.

Additional Information and Online Ordering

The Pierce RNA 3'-End Desthiobiotinylation Kit is optimized for labeling the 3'-end of single-stranded RNA using T4 RNA ligase. Once labeled, the RNA can be used as a probe or target for gel-shift EMSA reactions, Northern blots and protein-RNA interaction experiments. The desthiobiotinylated RNA also can be used to enrich for RNA binding proteins (RBP) using streptavidin affinity resin, because the desthiobiotin tag binds to streptavidin in a manner which allows gentle elution of the ribonucleoprotein complex. The labeling kit is also included as a component of the Thermo Scientific Pierce Magnetic RNA-Protein Pull-Down Kit.

Highlights:

  • Non-radioactive

    – a single biotin labels has similar detection sensitivity as radioactivity

  • Easy to use – RNA ligase and optimized reaction buffer are included

  • Economical – only a fraction of the cost purchasing synthetic biotinylated RNA probes

  • End-labeled – results in minimal disturbance of RNA secondary structure

  • Flexible

    – desthiobiotin label may be used for detection or as an affinity handle for streptavidin resin

Includes:

  • Desthiobiotinylated cytidine bisphosphate, T4 RNA ligase and buffer, unlabeled RNA oligonucleotide for use as a positive control, a biotinylated RNA probe standard, RNase inhibitor, glycogen, and ligation enhancing reagents

Recommended for:

  • Northern blotting
  • EMSA
  • Enrichment of RNA binding proteins (RBP)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 20 desthiobiotinylation reactions using 50 pmol of RNA each
• Desthiobiotinylated Cytidine Bisphosphate, 40 μL
• T4 RNA Ligase, 40 μL
• T4 RNA Ligase Reaction buffer (10X), 100 μL
• Non-labeled RNA Control, 100 μL
• Biotinylated IRE RNA Control, 35 μL
• RNase Inhibitor, 2 x 10 μL
• DMSO, 200 μL
• PEG (30%), 300 μL
• Glycogen, 20 μL
• Nuclease-free Water, 1.5 mL

Upon receipt, store at -20°C.
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 20
Product Line Pierce
Product Type RNA Labeling Kit
Quantity 20 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.