missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein-Free Blocking Buffer

Product Code. 10147483 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Chemical Name or Material:
Protein-Free (PBS)
Protein-Free (TBS)
Protein-Free T20 (PBS)
Protein-Free T20 (TBS)
Unit Size:
1 L
100mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material unitSize
10147483 1 L Protein-Free T20 (PBS) 1 L
13474299 100 mL Protein-Free (PBS) 100mL
10229663 1 L Protein-Free (PBS) 1 L
13484299 100 mL Protein-Free (TBS) 100mL
10269613 1 L Protein-Free (TBS) 1 L
10137483 1 L Protein-Free T20 (TBS) 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10147483 Supplier Thermo Scientific™ Supplier No. 37573

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background.

Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.

Features of Protein-Free Blocking Buffer include:

  • Protein-free blocker—minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
  • Applications—compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
  • Streptavidin-friendly—free of biotin; no interference with avidin-biotin detection systems
  • High-performance—optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)

Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Protein-Free T20 (PBS)
Concentration 1X
For Use With (Application) ELISA
Physical Form Liquid
Product Line Pierce
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.