Learn More
Thermo Scientific™ Pierce™ Protease and Phosphatase Inhibitor Tablets
Ensure complete protection from dephosphorylation and proteolytic degradation with these broad-spectrum protease and phosphatase inhibitor tablets.
£133.00 - £138.54
Specifications
Formulation | Inhibitors in proprietary excipient in amber glass vials |
---|---|
Form | Tablet |
Sufficient For | 200mL of sample (10mL each mini tablet) |
Quantity | 20 Tablets |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13393126
|
Thermo Scientific™
88668 |
Pierce Protease and Phosphatase Inhibitor Mini Tablets |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13313136
|
Thermo Scientific™
88669 |
Pierce Protease and Phosphatase Inhibitor Mini Tablets, EDTA Free |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation.
The Pierce Protease and Phosphatase Inhibitor Tablets contain aprotinin, bestatin, E-64 and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. The formulation is available with or without EDTA, a metalloproteinase inhibitor that may interfere with certain downstream applications. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.Highlights:
Complete All-in-one tablet contains both protease and phosphatase inhibitors
Easy-to-use the refrigerator-stable tablets are easily dissolved in buffers and lysis reagents
Two formulations available with or without EDTA
Compatible use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
Recommended for:
Protect intact, active cellular proteins from degradation and/or dephosphorylation by endogenous proteases and phosphatases; Preserve protein integrity and phosphorylation upstream of phosphoproteomic analyses and phospho-specific immunoassays
Specifications
Inhibitors in proprietary excipient in amber glass vials | |
Tablet | |
200mL of sample (10mL each mini tablet) | |
20 Tablets |