missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protease and Phosphatase Inhibitor Tablets

Product Code. 13393126 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Protease and Phosphatase Inhibitor
Protease and Phosphatase Inhibitor, EDTA Free
Unit Size:
20 tablets
This item is not returnable. View return policy

Product Code. 13393126

Brand: Thermo Scientific™ 88668

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ensure complete protection from dephosphorylation and proteolytic degradation with these broad-spectrum protease and phosphatase inhibitor tablets.

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation.

The Pierce Protease and Phosphatase Inhibitor Tablets contain aprotinin, bestatin, E-64 and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. The formulation is available with or without EDTA, a metalloproteinase inhibitor that may interfere with certain downstream applications. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:

  • Complete All-in-one tablet contains both protease and phosphatase inhibitors

  • Easy-to-use the refrigerator-stable tablets are easily dissolved in buffers and lysis reagents

  • Two formulations available with or without EDTA

  • Compatible use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays

Recommended for:

Protect intact, active cellular proteins from degradation and/or dephosphorylation by endogenous proteases and phosphatases; Preserve protein integrity and phosphorylation upstream of phosphoproteomic analyses and phospho-specific immunoassays

TRUSTED_SUSTAINABILITY

Specifications

Formulation Inhibitors in proprietary excipient in amber glass vials
Product Type Protease and Phosphatase Inhibitor
Form Tablet
Sufficient For 200mL of sample (10mL each mini tablet)
Quantity 20 Tablets
Composition Sodium fluoride, Sodium orthovanadate, Sodium pyrophosphate, beta-Glycerophosphate, Aprotinin, Bestatin, E-64, Leupeptin, EDTA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.