Learn More
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets,EDTA-free
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.
£21.85 - £21.93
Specifications
Includes | 1 vial of inhibitor tablets |
---|---|
Product Type | Protease and Phosphatase Inhibitor |
Content And Storage | 2°C to 8°C |
For Use With (Application) | protein purification, western blotting |
Sufficient For | 1 Tablet is Sufficient for 10 mL |
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
16316532
|
Thermo Scientific™
A32960 |
2 Tablets | Protease and Phosphatase Inhibitor |
£21.93
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
16326532
|
Thermo Scientific™
A32962 |
2 Tablets | Protease and Phosphatase Inhibitor |
£21.85
Each |
In Stock
Log in to see stock. |
||||
Description
The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.
Highlights:- Improved formulation—tablets dissolve quickly into a clear solution
- Economical – more cost-effective than other commercially available formulations for equivalent volumes
- Convenient—ready-to-use, fully disclosed, broad-spectrum formulations
- Compatible—use directly with Thermo Scientific™ Pierce™ BCA Assays
Specifications
1 vial of inhibitor tablets | |
2°C to 8°C | |
1 Tablet is Sufficient for 10 mL |
Protease and Phosphatase Inhibitor | |
protein purification, western blotting | |
2 Tablets |