Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Brand: Thermo Scientific™ A32965
213.34 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:Specifications
Protease Inhibitor Cocktail | |
Room Temperature | |
Pierce™ | |
Protease Inhibitor |
1 Vial, Store at 2°C to 8°C | |
Solid | |
50 mL Solution | |
20 Tablets |
Safety and Handling
- Pierce Protease Inhibitor Tablets, EDTA-Free
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
- P363-Wash contaminated clothing before reuse.
Supplemental information
- MIXTURE LIST-Contains : 4-(2-aminoethyl)-Benzenesulfonyl fluoride hydrochloride
For Research Use Only. Not for use in diagnostic procedures.