missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce Protease Inhibitor Tablets

Product Code. 13427766 Shop All Thermo Scientific Products
Click to view available options
Quantity:
20 Tablets
30 Tablets
Includes:
EDTA
EDTA Free
Unit Size:
20 tablets
30 tablets
This item is not returnable. View return policy

Product Code. 13427766

Brand: Thermo Scientific™ 88665

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ Pierce Protease Inhibitor Tablets and Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments.

Thermo Scientific™ Pierce Protease Inhibitor Tablets and Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments.

Additional Information and Online Ordering

Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A and are available in formulations with or without EDTA, a metalloproteinase inhibitor. The EDTA-free formulation has added compatibility with IMAC purification methods and 2D gel electrophoresis. Stable for storage at 4°C for up to 12 months, each large tablet is sufficient for 50mL of solution and each mini tablet is sufficient for 10mL of solution. Both sizes and formulations of tablet are suitable for reagent preparation prior to protein extraction from tissue and cultured cells.

Highlights:

  • Economical −both large and mini tablets are most cost effective than other commercially available formulations for equivalent volumes
  • Easy−to−use − the refrigerator−stable tablets are easily dissolved in buffers and lysis reagents
  • Two formulation − available with or without EDTA
  • Compatible − use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
TRUSTED_SUSTAINABILITY

Specifications

Formulation Inhibitors in proprietary excipient in amber glass vials
Includes EDTA
Product Type Protease Inhibitor Mini Tablets
Form Tablet
Product Line Pierce™
Sufficient For 300mL of sample (10mL each mini tablet)
Quantity 30 Tablets
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.