missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dulbecco's Modified Eagle's Limiting Medium (DMEM-LM)

Product Code. 10425145 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10425145 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10425145 Supplier Thermo Scientific™ Supplier No. 30030
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diazirine analogs of leucine and methionine to express proteins in cell culture that will crosslink their protein interactors upon UV-light activation in vivo.

Thermo Scientific™ Pierce Photoreactive Amino Acids are analogs of L-Leucine and L-Methionine that can be incorporated into proteins during synthesis and then light-activated to covalently crosslink interacting proteins in cells.

Thermo Scientific L-Photo-Leucine and L-Photo-Methionine are analogs of L-Leucine and L-Methionine amino acids that have activatable diazirine side chains capable of chemical crosslinking to adjacent molecules when exposed to ultraviolet light. Like their naturally occurring counterparts, these photo-reactive amino acids can be endogenously incorporated into the primary sequence of proteins during synthesis. Protein-protein interaction domains involving the synthesized proteins in their native environments can then be covalently crosslinked by activating the diazirine groups. Traditional crosslinking experiments require adding bifunctional chemical crosslinkers and associated reagent solvents, which can adversely affect the cell biology being studied. Using photo-reactive amino acids minimizes these potentially adverse effects and enables both stable and transient protein interactions in cells to be studied and characterized with a high degree of confidence.When used in combination with specially formulated limiting cell media that is devoid of leucine and methionine, the photo-activatable derivatives are treated like naturally occurring amino acids by the protein synthesis machinery. As a result, they can be substituted for leucine or methionine in the primary sequence of proteins. When exposed to UV light the diazirine rings become reactive intermediates that form covalent bonds with nearby protein side chains and backbones. Naturally associating binding partners are then instantly trapped. Crosslinked protein complexes are detected by decreased mobility on SDS-PAGE followed by Western blot detection, size-exclusion chromatography, sucrose density-gradient sedimentation or mass spectrometry.

Highlights:

  • In vivo labeling – incorporate photo-reactive group into proteins using normal cellular machinery

  • In vivo crosslinking – find interacting proteins in the native cellular environment

  • Increased specificity compared to traditional methods – crosslink interacting proteins correctly positioned at their interfaces within protein interaction domains

  • Complementary to traditional methods – provides detailed characterization of protein interactions when compared to results with formaldehyde and other more general crosslinking approaches

  • Efficient recovery – 90% protein recovery in cell lysates after crosslinking

  • Compatible – crosslink proteins expressed in a wide variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3 and C6

  • Easy to use – reagents are photo-stable under normal laboratory lighting, eliminating the need to work in the dark

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Crosslinker
Product Line Pierce
Quantity 500 mL
Content And Storage Upon receipt store at 4°C.
Description Pierce Dulbecco's Modified Eagle's Limiting Medium (DMEM-LM)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.