missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Phosphatase Inhibitor Mini Tablets

Product Code. 16306532 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Tablets
20 Tablets
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16306532 2 Tablets Each
15691759 20 Tablets Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.
Product Code. 16306532 Supplier Thermo Scientific™ Supplier No. A32958
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation with a broad spectrum phosphatase inhibitor formulation.

Pierce Phosphatase Inhibitor Mini Tablets protect phosphoproteins from serine/threonine phosphatases and tyrosine phosphatases. Each Phosphatase Inhibitor Tablet contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate for broad-spectrum inhibition of phosphatase activity. Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution. The Pierce Phosphatase Inhibitor Tablets are suitable for use with any cell or tissue extract to preserve protein phosphorylation for downstream applications.

Highlights:
  • Economical&mdsashmore cost effective than other commercially available formulations
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Complete&mdsashthe tablets inhibit acid, alkaline, and protein phosphatases
  • Compatible&mdsashuse with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
TRUSTED_SUSTAINABILITY

Specifications

Includes 1 vial of inhibitor tablets
Content And Storage 2°C to 8°C
For Use With (Application) protein purification, western blotting
Sufficient For 1 Tablet is Sufficient for 10 mL
Quantity 2 Tablets

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.