missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Pierce™ Peptide Retention Time Calibration Mixture
Click to view available options
Quantity:
50 μL
Unit Size:
Pack of 850
Description
- Single use eliminates contamination from carry over or absorption
- Excellent for sensitive batch work or low speed centrifugation
- Color coded caps available separately
Specifications
Specifications
| Description | 1000 x g |
| Capacity (Metric) | 15 mL |
| Length (Metric) | 150 mm |
| Material | Polystyrene |
| Disposable | Single-use |
| Quantity | 50 μL |
| Diameter (Metric) | 16 mm |
| Product Type | Test Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction