Learn More
Thermo Scientific™ TMS(PEG)12 Branched Methyl-PEG-NHS-Ester Reagent
Shop All Thermo Scientific ProductsDescription
A three-branched, polyethylene glycol compound (3 times 12 PEG units) activated as an NHS ester for covalent PEGylation of primary amines on proteins or assay surfaces.
Thermo Scientific™ Pierce TMS(PEG)12 is a three-branched, polyethylene glycol compound (3 times 12 PEG units) activated as an NHS ester for covalent pegylation of primary amines (e.g., lysines) on proteins or assay surfaces.
TMS(PEG)12 is the abbreviation for a branched trimethyl (TM) and succinimide ester (S) derivative of polyethylene glycol (PEG) for efficient and specific modification of primary amines. Each methyl-terminated PEG (mPEG) branch contains 12 ethylene glycol units. The three branches are attached to a 4-unit PEG stem that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at the distal end. The NHS ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.
Highlights:
- NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
- Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Easy-to-follow instructions increase the likelihood of a successful outcome
Specifications
Specifications
| Chemical Reactivity | Amine |
| Content And Storage | Store desiccated at -20°C. |
| Label Type | Polyethylene Glycol (PEG), Blocking Groups |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Spacer | 12 ethylene glycol units |
| Form | Solid |
| Quantity | 1 g |
| Solubility | Water |
| Product Type | NHS-Ester Reagent |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.