missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DHB MALDI Matrix, Single-Use

Product Code. 10046153 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
24 x 4 mg
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10046153 24 x 4 mg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046153 Supplier Thermo Scientific™ Supplier No. 90033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).

Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.

The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.

Highlights:

  • Highly purified, recrystallized reagent
  • Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
  • Provides accurate quantity of matrix every time
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce DHB MALDI Matrices
For Use With (Equipment) Mass Spectrometer
Product Type MALDI Matrix
Content And Storage Upon receipt store product at 4°C.
Starting Material Small Molecules
Final Product Type Small Molecules
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 24 x 4 mg
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.