Learn More
Thermo Scientific™ Pierce™ LDH Cytotoxicity Assay Kit
Quantitatively measure lactose dehydrogenase released into the media from damaged cells as a biomarker for cellular cytotoxicity and cytolysis using a colorimetric assay.
Brand: Thermo Scientific™ 88954
Code : X3
Additional Details : Weight : 0.45360kg
Description
Quantitatively measure lactose dehydrogenase (LDH) released into the media from damaged cells as a biomarker for cellular cytotoxicity and cytolysis using a colorimetric assay with Thermo Scientific™ Pierce LDH Cytotoxicity Assay Kit.
Additional Information and Online Ordering »
Highlights:
Convenient – add–mix–read assay format for adherent and suspension cells
Colorimetric – quantitatively measures LDH release by formation of colored product
Robust – uses stable LDH enzyme activity as a cytotoxic marker
Flexible – ideal for high–throughput screening
Non–radioactive – safe alternative to 51Cr–release cytotoxicity assays
Includes:
Kit contains substrate mix, assay buffer, 10X lysis buffer, stop solution, and LDH positive control
Requires:
Microplate reader capable of reading absorbance at 490nm and 680nm, flat-bottom clear 96–well plate compatible with spectrophotometer, multichannel pipette
Recommended for:
- Measure in vitro cytotoxicity mediated by chemicals, immune cells, or siRNA or microRNA
- Detect cytolysis in bioreactor or 3D tissue engineering applications
Specifications
Pierce LDH Cytotoxicity Assay Kit | |
1000 Reaction Kit | |
10X Lysis Buffer, 12mL, Substrate Mix, 5 x lyophilizate, Stop Solution, 60mL, LDH Positive Control, 30µL, Assay Buffer, 5 x 0.6mL | |
1000 reactions in 96-well plates or 4000 reactions in 384-well plates |