missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ LAL Chromogenic Endotoxin Quantitation Kit

Product Code. 12117850 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 12117850

Brand: Thermo Scientific™ 88282

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Accurately measure endotoxin levels (LPS, lipopolysaccharides) in protein, peptide or antibody samples using our Limulus Amebocyte Lysate (LAL) assay.

The Thermo Scientific™ Pierce LAL Chromogenic Endotoxin Quantitation Kit measures the amount of endotoxin in a protein, peptide or antibody sample using the Limulus Amebocyte Lysate (LAL) assay.

The endotoxin concentration in a sample is measured using the Pierce LAL Chromogenic Endotoxin Quantitation Kit via a chromogenic signal generated in the presence of endotoxins. Samples can be measured on a microplate absorbance reader at 405nm. A standard curve is created using the E. coli endotoxin standard included with each kit to calculate endotoxin levels as low as 0.1 EU/mL. Protein and antibody samples can be assayed in as little as 30 minutes. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation and/or sepsis in tissue culture cells and animals injected with endotoxin-contaminated proteins.

Highlights:

  • Sensitive—detect as little as 0.1 EU/mL

  • Fast—perform this assay in 30 minutes

  • Economical—assay requires only 10μL of a protein sample

  • AccurateE. coli O111:B4 standard in each kit enables accurate endotoxin quantitation

  • Versatile—405nm absorbance reading is compatible with common ELISA plate readers

Recommended for:

Quantitation of endotoxin levels in a protein, peptide or antibody solution

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack.
Assay [other]
Detection Method Colorimetric
For Use With (Equipment) Spectrophotometer, Microplate Reader
Product Line Pierce
Quantity 50 Tests
For Use With (Application) Solution-Based Detection, Absorbance
Target Not Target-Specific
Type Endotoxin Quantitation Kit
Format Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado