Learn More
Invitrogen™ In-Cell ELISA Kit, Colorimetric
Description
The In-Cell ELISA Colorimetric Detection Kit is a simple method for quantifying intracellular proteins in whole cells. Principle of the method To perform the assay, cells are first plated, treated and fixed. Expression of the protein(s) of interest is monitored in wells of a microplate using target-specific primary antibodies (see the Important Product Information section for antibodies included in each kit) and a horseradish peroxidase (HRP)-conjugated detection reagent. The kit is supplied with a whole-cell stain to control for differences in cell plating, which is important when measuring relative levels of a protein with different treatments or assessing its post-translational modification (PTM) form. After staining, the results are analyzed by normalizing the absorbance (HRP activity) values to cell number, which adjusts for the cell plating differences among the wells. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Highlights:
- Two detection formats available: colorimetric (for standard plate readers) and near infrared fluorescent (for infrared imaging systems)
- Simple,fast assay format and the ability to perform multiplex experiments
- No special sample preparation required
- Higher throughput compared with Western blotting
- Cost efficient (one well vs. one lane) compared to Western blotting
Specifications
Specifications
| Description | Pierce In-Cell ELISA Colorimetric Detection Kit |
| Product Type | ELISA Kit |
| Quantity | 4 x 96 Tests |
| Conjugate | HRP |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Sample Type | Supernatant |
| Sample Volume | Supernatant 10,000 cells |
| Test Time | 1 hr. 20 min. |
| Total Assay Time | 24 hr. |
| Kit Contents | Blocking Buffer, 20X Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, HRP Conjugate, TMB Substrate, TMB Stop Solution, Janus Green Whole-Cell Stain, Elution Buffer, Thin Plate Seal Assembly, Components included only in the target-specific kits:Antibody #1 and Antibody #2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.