Learn More
Thermo Scientific™ Pierce™ NeutrAvidin™ Coated High Sensitivity Plates, Clear, 8-Well Strip
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce High Sensitivity NeutrAvidin Plates are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.
Highlights:
Sensitive – detect down to 5ng/mL of biotinylated IgG
Specific – low background with high signal-to-noise ratios
Versatile – use either fluorescence, chemiluminescence or colorimetric detection
Robust – broad dynamic range
Ready to use – plates are supplied pre-blocked to save time
Specifications
Specifications
| Binding Property | High Sensitivity, Affinity |
| Content And Storage | Upon receipt store at 4°C. |
| Description | Pierce NeutrAvidin Coated High Sensitivity Plates, Clear, 96-Well Strip |
| Detection Method | Chemiluminescence |
| Packaging | 5 Plate |
| Product Line | NeutrAvidin Pierce |
| Color | Clear |
| Product Type | Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.