missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ HeLa Digest/PRTC Standard

Product Code. 17175802 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 10 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17175802 5 x 10 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 17175802 Supplier Thermo Scientific™ Supplier No. A47997
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

The Thermo Scientific Pierce HeLa Digest/PRTC Standard is a validated complex mammalian protein digest containing additional PRTC heavy isotope-labeled peptides designed for LC/MS performance testing.

The Thermo Scientific Pierce HeLa Digest/PRTC Standard is a validated complex mammalian protein digest containing additional PRTC heavy isotope-labeled peptides designed for LC/MS performance testing.

Features of HeLa Digest/PRTC Standard include:

  • Convenient—pre-mixed formulation of HeLa digest and PRTC standard in a ready-to-use format
  • Informative—enables robust performance assessment of LC/MS instruments
  • Verified—high-quality, consistent mixture documented via lot-specific certificates of analysis
  • Stable—provided in a lyophilized format

The Pierce HeLa Digest/PRTC Standard is a lyophilized mixture of 10 μg of HeLa protein digest with 5 pmol PRTC peptides to assess chromatography MS instrument performance. The protein digest is derived from a well-established adenocarcinoma reference HeLa S3 cell line that expresses over 15,000 proteins with relevant post-translational modifications. This complex digest is an ideal standard for complex proteome mass spectrometry applications.

The included Pierce Peptide Retention Time Calibration Mixture contains 15 synthetic heavy isotope-labeled peptides mixed at an equimolar ratio that elute across the chromatographic gradient. The observed retention times and hydrophobicity factors (HFs) for these calibrant peptides are fit to a linear equation to determine the slope of the retention time/HF relationship. This equation and the HF of an uncharacterized peptide are then used to predict retention time. In combination, the mixture is a powerful tool to optimize and assess LC parameters, identify the total peptide elution window, and optimize and monitor MS instrumentation.

This standard is specifically formulated for LC/MS experiments and does not contain salts or detergents. Using the standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.

Applications:

  • Qualitative LC assessment
  • QC assay tool for assessment of LC/MS system performance
  • LC/MS standardization
  • LC/MS method development and optimization
  • Normalize results for variation in retention times and peak intensities between runs
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HeLa Digest/PRTC Standard
Packaging Lyophilized in glass vial
Synonym HeLa/PRTC
Quantity 5 x 10 μg
Grade MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.