missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Goat Anti-Rabbit IgG Antibody Coated Plates, Clear, 96-Well

Product Code. 10723944 Shop All Thermo Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 10723944

Brand: Thermo Scientific™ 15135

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.

Thermo Scientific™ Pierce Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.

These plates are pre-coated with goat anti-mouse (GAM) or goat anti-rabbit (GAR) IgG polyclonal antibodies, allowing assay-specific antibodies to be captured even from impure protein samples. The plates are supplied pre-blocked and ready-to-use. Pierce Secondary Antibody Coated Plates can be used for direct, indirect, competitive or sandwich assays and are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.

Highlights:

  • Prevents antibody denaturation caused by direct binding
  • Unlike Protein A or G, these bind only to target species
  • Antibody-binding capacity is higher than direct adsorption
  • Pre-blocked to reduce nonspecific binding
  • Less expensive than Protein A or G plates
TRUSTED_SUSTAINABILITY

Specifications

Binding Property Standard Capacity, Affinity
Content And Storage Note: Plates are activated to 100 μL and supplied pre-blocked with SuperBlock™ Blocking Buffer.

Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag and store desiccated at 4°C.
Detection Method Colorimetric
Packaging 5 Plates
Product Line Pierce
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 96
Surface Treatment Antibody
Plate Blocking SuperBlock™ Buffer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.