Learn More
Description
Thermo Scientific™ Pierce Glutathione Magnetic Beads are superior GST-binding particles for automated magnetic separations.
Additional Information and Online Ordering
Highlights:
High-performance beads – non-aggregating, silanized, iron oxide, paramagnetic microparticles provide exceptional uniformity for automated HTS applications
Stable affinity ligand – glutathione is covalently immobilized to particles, ensuring leach-resistance and clean purification products
High capacity – binding capacity is sufficient for both routine and demanding magnetic separation procedures
GST-tag affinity – standard glutathione-GST affinity purification chemistry in a magnetic format; bind in physiologic buffers and elute with 50mM reduced glutathione (prepared with Part No. 78259)
Properties of Glutathione Magnetic Beads:
- Composition: iron oxide particles with reduced glutathione (GSH) covalently attached to the surface and encapsulated by crosslinked agarose
- Magnetization: superparamagnetic (no magnetic memory)
- Bead size: 1 to 10μm (mean diameter)
- Concentration: 25% slurry (based on volume) in 20% ethanol
- Binding capacity: 5 to 10mg of GST fusion protein per milliliter of settled beads (i.e., 4mL of 25% slurry)
Requires:
Thermo Scientific KingFisher 96 or KingFisher Flex Instrument or other magnetic separator device
Specifications
Specifications
| Description | Pierce Glutathione Magnetic Beads |
| Formulation | Glutathione on paramagnetic iron oxide particles (1 to 10um dia.) slurried in 20% ethyl alcohol |
| Support Type | Magnetic Bead |
| Quantity | 4 mL |
| Sufficient For | Binding 5 to 10mg GST per mL of beads |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.