missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers

Product Code. 31924099 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 4 mL
Unit Size:
8mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31924099 2 x 4 mL 8mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31924099 Supplier Thermo Scientific™ Supplier No. A51739
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A room-temperature stable, ready-to-use liquid formulation for the calibration of Thermo Scientific™ Mass Spectrometers with the integrated Auto-Ready ion source.

  • Extended mass range improves mass accuracy
  • Positive and negative mode calibration, a single formulation with extended mass ranges for both ionization calibration modes
  • High-quality, rigorously-tested formulation with lot-specific certificates of analysis
  • Bottle may be directly loaded into the instrument for scheduled automatic calibration
  • Stable at room temperature for 18 months, and within the auto-ready mass spectrometer for up to 4 months
TRUSTED_SUSTAINABILITY

Spécification

Physical Form Liquid
Color Colorless
Quantity 2 x 4 mL
Percent Purity Mixture
Grade LC-MS grade

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.