Learn More
Description
The Thermo Scientific™ Pierce Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.
Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit contains reagents to measure the activity of firefly luciferase in mammalian whole cell lysates. Containing a convenient lysis buffer, assay buffer and firefly substrate, this kit provides a complete reagent system for the intracellular detection of promoter or pathway activity. Light output resulting from the luciferase reaction is captured using a luminometer and can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression. These reagents are optimized to work with Thermo Scientific Firefly Luc Plasmids; however, the Flash Assay Kit may be used to detect the activity of other firefly or ATP-dependent luciferases utilizing D-luciferin as a substrate.
Highlights
- Highly sensitive detection of firefly luciferase activity
- Use with Pierce Firefly Luciferase Signal Enhancer for best results (maximum signal)
- Working solution stable for up to 2 months at -20°C
- Assay reagents compatible with other firefly luciferases
- Amenable to automation
- Contains a universal cell lysis buffer and optimized flash assay reagent
- Safe non-radioactive assay
Requires
Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; For best results (maximum signal), use with Thermo Scientific Pierce Firefly Luciferase Signal Enhancer
Recommended applications
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway/protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Firefly Luciferase |
| Quantity | 1000 Reaction Kit |
| Content And Storage | • Firefly Flash Assay Buffer, 5 mL, store at 4°C • D-Luciferin, 3 mg, store at 4°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature Upon receipt store kit at 4°C or store individual components as indicated above. |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Label Type | Enzyme Labeled |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.