missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Endotoxin-Free Magnesium Chloride (MgCl2) (10 mM)
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
Pack of 5
Description
Pierce Endotoxin-Free MgCl2 is suitable for the preparation of samples for endotoxin detection (LAL) assay. As a diluent, it will eliminate the inhibitory chelation effects of EDTA. It contains <0.005 EU/mL endotoxin.
Specifications
Specifications
| Description | Endotoxin-Free MgCl2 buffer |
| Concentration | 10 mM |
| For Use With (Equipment) | LAL Endotoxin Assay |
| Quantity | 5 x 10 mL |
| Packaging | Vial |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction