missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Biotin 3' End DNA Labeling Kit

Product Code. 10036813 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10036813 20 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10036813 Supplier Thermo Scientific™ Supplier No. 89818
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotinylate single-stranded DNA primers for non-radioactive EMSA and other nucleic acid detection methods.

The Thermo Scientific Pierce Biotin 3' End DNA Labeling Kit is for tagging single-stranded DNA primers with biotin for use in non-radioactive electrophoretic mobility shift assays (EMSA) and other nucleic acid detection methods.

The DNA biotinylation procedure uses terminal deoxynucleotidyl transferase (TdT) to catalyze nontemplate-directed nucleotide incorporation onto the 3'-OH end of single-stranded DNA. TdT exhibits a substrate preference of single-stranded DNA, but it will label duplex DNA with 3' overhangs and blunt duplexes, albeit with a lower efficiency.

Features of the Biotin 3' End DNA Labeling Kit:

  • Non-isotopic labeling eliminates the hassle of hazardous radioactive materials or difficult-to-dispose-of waste
  • 1-3 biotinylated ribonucleotides onto the 3' end of DNA strands for less interference with hybridization or sequence-specific binding of proteins
  • Biotin-labeled probes are stable for more than one year
  • 30-minute labeling procedure is fast and efficient

    The Biotin 3' End DNA Labeling Kit has been optimized to incorporate 1-3 biotinylated ribonucleotides (Biotin-11-UTP) onto the 3' end of DNA strands. This labeling strategy has the advantage of localizing the biotin to the 3' end of the probe where it will be less likely to interfere with hybridization or sequence-specific binding of proteins. Biotin-labeled DNA probes can be used to facilitate non-isotopic detection in a variety of applications including electrophoretic mobility shift assays (EMSA), Northern or Southern blots, colony hybridizations or in situ hybridizations.

  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Sufficient For: 20 labeling reactions (each with 5 pmol DNA)
    • 1 mL 5X TdT Reaction Buffer (-20°C)
    • 5 μL Terminal Deoxynucleotidyl Tranferase (-20°C)
    • 1 μL Biotin-11-UTP (≤4°C)
    • 14 μL Unlabeled Control Oligo (-20°C)
    • 4 μL Biotin-Control Oligo (-20°C)
    Format Kit
    Labeling Target DNA
    No. of Reactions 20
    Product Line Pierce
    Product Type DNA Labeling Kit
    Quantity 20 Reactions
    compliance-icons compliance-icons compliance-icons
    Product Identifier
    • Pierce Biotin 3' End DNA Labeling Kit
    Signal Word
    • Danger
    Hazard Category
    • Acute toxicity Category 4
    • LONG-TERM AQUATIC HAZARD Chronic 2
    • Carcinogenicity Category 1B
    Hazard Statement
    • H302-Harmful if swallowed.
    • H350-May cause cancer.
    • H411-Toxic to aquatic life with long lasting effects.
    Precautionary Statement
    • P201-Obtain special instructions before use.
    • P273-Avoid release to the environment.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
    • P308+P313-IF exposed or concerned: Get medical advice/attention.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contain: Cobalt(II) chloride,hexahydrate

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.