Learn More
Description
Thermo Scientific™ Pierce DAB and the DAB Substrate Kit enable chromogenic detection of horseradish peroxidase (HRP) activity based the action of 3,3'-diaminobenzidine (DAB) in blotting and tissue staining methods.
Highlights:
HRP substrate – for detection of horseradish peroxidase activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
Chromogenic – no special equipment needed for visualization; the DAB reaction produces brown bands or spots at sites of reaction with HRP-conjugated antibodies or probes
Package options – choose powder for custom formulation with buffer and peroxide or a complete kit of preformulated solutions for immediate use
Specifications
Specifications
| Description | Pierce DAB Powder |
| Detection Method | Chromogenic |
| Quantity | 10 g |
| Conjugate | HRP |
| Sufficient For | 2000 uses at 1X (0.5mg/mL) and 10mL each |
| Kit Contents | Single component |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.