missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt
Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.
£111.00 - £682.00
Specifications
Content And Storage | Store at or below -20°C. |
---|---|
Detection Method | Bioluminescence |
For Use With (Application) | ATP Assay, Gene Reporter Assay |
Product Line | Pierce™ |
Substrate Properties | Chemical Substrate |
Product Code | Brand | Quantity | Salt Form | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Salt Form | Price | Quantity & Availability | ||||
15285723
|
Thermo Scientific™
88291 |
100 mg | Monosodium Salt |
£111.00
100mg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15225733
|
Thermo Scientific™
88294 |
1 g | Monopotassium Salt |
£682.00
1g |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.
We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.Specifications
Store at or below -20°C. | |
ATP Assay, Gene Reporter Assay | |
Chemical Substrate |
Bioluminescence | |
Pierce™ | |
Luciferase Substrate |
For Research Use Only. Not for use in diagnostic procedures.