missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 3 (Cleaved) Multispecies In-Cell ELISA Kit, Colorimetric

Product Code. 11598120
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11598120 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11598120 Supplier Invitrogen™ Supplier No. 62218

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In-cell ELISA Kit

The Cleaved Caspase 3 Multispecies In-Cell ELISA Colorimetric Detection Kit is a simple method for quantifying intracellular proteins in whole cells. Principle of the method To perform the assay, cells are first plated, treated and fixed. Expression of the protein(s) of interest is monitored in wells of a microplate using target-specific primary antibodies (see the Important Product Information section for antibodies included in each kit) and a horseradish peroxidase (HRP)-conjugated detection reagent. The kit is supplied with a whole-cell stain to control for differences in cell plating, which is important when measuring relative levels of a protein with different treatments or assessing its post-translational modification (PTM) form. After staining, the results are analyzed by normalizing the absorbance (HRP activity) values to cell number, which adjusts for the cell plating differences among the wells. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits,large and small,that dimerize to form the active enzyme. This protein cleaves and activates caspases 6,7 and 9,and the protein itself is processed by caspases 8,9 and 10. It is the predominant caspase involved in the cleavage of amyloid-beta 4A precursor protein,which is associated with neuronal death in Alzheimer′s disease. Alternative splicing of this gene results in two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42574
Description Pierce Cleaved Caspase 3 Colorimetric In-Cell ELISA Kit
Conjugate HRP
Immunoassay Kit Format Multi-component kit with HRP-conjugate
Product Type ELISA Kit
Sample Type Supernatant
Sufficient For 96 microplate wells of overnight-cultured cells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 836
Gene Symbol CPP32,CPP32B,SCA-1
Kit Contents Blocking Buffer, 20X Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, HRP Conjugate, TMB Substrate, TMB Stop Solution, Janus Green Whole-Cell Stain, Elution Buffer, Thin Plate Seal Assembly, Components included only in the target-specific kits:Antibody #1 and Antibody #2, TMB Substrate, 58mL
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Supernatant, 10,000 cells
Storage Requirements 2°C to 8°C
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 24 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.