missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ C-18 Tips

Product Code. 10506495 Shop All Thermo Scientific Products
Click to view available options
Quantity:
8 Tips
Unit Size:
Pack of 8
This item is not returnable. View return policy

Product Code. 10506495

Brand: Thermo Scientific™ 87783

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Capture, concentrate and desalt 10 to 100 microliter samples of peptides and proteins for MALDI mass spectrometry with these ready-to-use pipette-tip columns of C18 resin.

Thermo Scientific™ Pierce C-18 Tips are ready-to-use pipette-tip columns of C18 resin that enable fast and efficient capture, concentration, desalting and elution of peptides and proteins for MALDI mass spectrometry and other methods.

Improve protein analysis results with Pierce C18 Tips by removing urea, salts and other contaminants before MS analysis. The tips are ideal for preparing samples for matrix-assisted laser desorption ionization (MALDI) or nanoelectrospray ionization techniques. The Pierce C18 Tips are available in convenient 10 and 100μL tips with binding capacities of 8 and 80μg, respectively.

Highlights:

  • Monolithic C18 fast-flow tips have low resistance and improved flow characteristics compared to other commercially available tips
  • High binding capacity in a convenient tip format
  • Two pipette tip formats for processing up to 8 or 80μg of sample
  • Eight packs and 96-tip trays are automation compatible
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce C-18 Tips, 100uL bed
Formulation Pipette tips containing 100µL of porous C-18 reversed-phase resin
Standard Concentration 98 to 100%
Sufficient For Processing 8 samples (100µL each) containing up to 80µg peptide each
Content And Storage Room Temperature
Quantity 8 Tips
For Use With (Application) MALDI-MS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.