missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Bovine Gamma Globulin Standard Pre-Diluted Set

Product Code. 10619034 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
7 x 3.5 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10619034 7 x 3.5 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10619034 Supplier Thermo Scientific™ Supplier No. 23213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality, calibrated BGG solutions (2mg/mL) to serially dilute to create protein assay standard curves to measure antibody (IgG) concentration.

Thermo Scientific™ Pierce BGG Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.

These bovine gamma globulin (BGG) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BGG is an accepted reference protein for total protein quantitation of purified antibodies or immunoglobulin-rich samples. The IgG standard is precisely formulated at 2mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution of purified BGG (Fraction II) is calibrated by direct comparison to an internal standard to ensure lot-to-lot consistency and accuracy. The product is available in two convenient package formats: 1mL glass ampules and a set containing seven ready-to-use dilutions (3.5mL each).

Highlights:

  • Antibody standard – best reference standard for immunoglobulin quantitation in colorimetric protein assays

  • Bradford standard – best general protein standard in coomassie-based (Bradford) protein assays

  • Pure and stable – supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable

  • Accurate and consistent – precisely formulated at 2.00 +/-0.03mg/mL, ensuring excellent lot-to-lot consistency

  • Convenient – choose a package of 1mL ampules or a complete set containing seven ready-to-use dilutions

Includes:

2mg/mL BGG solution (Part No. 23212) or 7-solution set of different concentrations (Part No. 23213)

Recommended for:

Protein assay immunoglobulin quantitation standard; Antibody recovery control for desalting and other column procedures; Loading control for SDS-PAGE; General calibration of spectrophotometer UV-lamp (A280)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage BGG in 0.9% sodium chloride with sodium azide; one each of the following μg/mL concentrations: 125, 250, 500, 750, 1000, 1500, 2000

Store at 2–8°C.
Description Pierce Bovine Gamma Globulin Standard Pre-Diluted Set
Assay Protein Standards
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
For Use With (Application) Internal Standard
Quantity 7 x 3.5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.