missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ BCA Protein Assay Reagent A

Product Code. 10198543 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3.75 L
Unit Size:
1000mL
250mL
3.75 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10198543 250 mL 250mL
10752735 3.75 L 3.75 L
11881345 1 L 1000mL
10056623 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10198543 Supplier Thermo Scientific™ Supplier No. 23221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Estimate visually, or measure with a standard spectrophotometer or plate reader at 562 nm.

Thermo Scientific™ Pierce BCA Protein Assay Kit remains one of the most popular protein quantitation methods worldwide.

Additional Information and Online Ordering

  • A component of the Pierce BCA Protein Assay Kit
  • Kit is a two-component, high-precision, detergent-compatible assay that is used for total protein concentration determination compared to a protein standard
  • Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research
  • The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications
  • Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy
  • Estimate visually or measure with a standard spectrophotometer or plate reader at 562 nm
  • Exhibits less protein-to-protein variation than dye-binding methods (Bradford)
  • Unaffected by typical concentrations of most ionic and non-ionic detergents
  • Assay has a 30 min. incubation; much easier and four times faster than classical Lowry methods
  • Linear working range for BSA of 20 to 2000 μg/mL
  • Detect down to 5 μg/mL with the enhanced protocol
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Description Pierce BCA Protein Assay Reagent A
Assay BCA Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 250 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.