Learn More
Invitrogen™ Pierce™ Avidin, Fluorescein (FITC) Conjugated
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Avidin and Conjugates include purified avidin protein from hen egg whites, plus avidin-HRP, avidin-AP and other conjugated forms for use in biotin-based detection methods.
Highlights:
Native avidin – glycoprotein from chicken egg whites, 67kDa, pI equals 10
Soluble – glycosylation and isoelectric point renders avidin more soluble than streptavidin
Affordable – significantly less expensive than recombinant streptavidin
ABC staining – avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
Pure protein and conjugates – choose pure, unconjugated avidin for use in ABC staining or peroxidase or alkaline phosphatase forms for membrane and plate-based assays
Specifications
Specifications
| Concentration | 5.0 mg/mL |
| Conjugate | FITC (Fluorescein) |
| Quantity | 5 mg |
| Content And Storage | Store at 2–8°C. Shipped at ambient temperature. |
| Form | Liquid |
| Product Line | Pierce |
| Product Type | Avidin Conjugate (fluorescent) |
| Target Molecule | Biotin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.