Learn More
Thermo Scientific™ Pierce™ Anti-GST Coated Plates, Clear, 8-Well Strip
Detect glutathione S-transferase (GST)-tagged proteins with these ready-to-use pre-blocked, mouse monoclonal anti-GST antibody coated polystyrene microplates.
Brand: Thermo Scientific™ 15144
Code : PN
Additional Details : CAS Number : 1141-06-6 Weight : 0.10000kg
Description
Thermo Scientific™ Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
Additional Information and Online Ordering »
Highlights:
Antibody-based – plates are coated with mouse monoclonal anti-GST antibody
Sensitive – detects GST fusion proteins down to 7.5ng/well
Flexibility – binds native or denatured forms of GST; an alternative to glutathione-coated plates
Ready to use – pre-blocked with SuperBlock Blocking Buffer
Compatibility – Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion protein binding
Superior performance – improved signal-to-noise ratio relative to competitor plates
Specifications
Standard Capacity, Affinity | |
Colorimetric | |
Pierce | |
Detection Plate, Immunoassay, ELISA | |
96 | |
SuperBlock™ Buffer |
Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature. | |
5 Plate | |
Clear | |
Polystyrene | |
Antibody |
For Research Use Only. Not for use in diagnostic procedures.