missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKT Multispecies In-Cell ELISA Kit, Near Infrared

Product Code. 11568100
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11568100 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11568100 Supplier Invitrogen™ Supplier No. 62220

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In-cell ELISA Kit

The AKT Multispecies In-Cell ELISA Near Infrared Detection Kits provide a simple and convenient method for quantifying intracellular proteins in whole cells. Principle of the method This kit enables simultaneous detection of post-translational-modified protein (PTM) and the corresponding unmodified protein, or two different targets within the same well. Simultaneously detecting targets within an ELISA well eliminates variability caused by differences in cell plating. The expression levels of the protein(s) are monitored using primary antibodies specific to the targets and corresponding species-specific near-infrared Thermo Scientific DyLight-Conjugated Secondary Antibodies. Relative protein activation or inactivation is determined as a ratio of modified protein levels to the corresponding unmodified protein measured using a modification-specific antibody and an antibody to unmodified protein, respectively. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

AKT (protein kinase B, PKB, RAS-alpha) is a 57kDa serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. AKT is activated by insulin, PI3K, IGF1 and various other growth and survival factors. AKT promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. There are three mammalian isoforms of Akt: AKT1 (PKB alpha), AKT2 (PKB beta) and AKT3 (PKB gamma) with AKT2 and AKT3 being approximately 82% identical with the AKT1 isoform. Each isoform has a pleckstrin homology (PH) domain, a kinase domain and a carboxy terminal regulatory domain. AKT was originally cloned from the retrovirus AKT8, and is a key regulator of many signal transduction pathways. The AKT pathway is a major target for cancer therpeutics where AKT signaling dysfunction has implicated in many types of cancer, including cancer syndromes known as phakomatoses.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31749, P31751, Q9Y243
Description Pierce AKT Near Infrared In-Cell ELISA Kit
Conjugate Unconjugated
Immunoassay Kit Format Multi-component kit with fluorescent conjugates
Product Type ELISA Kit
Sample Type Supernatant
Sufficient For 96 microplate wells of overnight-cultured cells
Target Species Human
For Use With (Equipment) Fluorescent Microplate Reader
Gene ID (Entrez) 10000,207,208
Gene Symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,HIHGHH,PKBB,PKBBETA,PRKBB,RAC-BETA,MPPH,MPPH2,PKB-GAMMA,PKBG,PRKBG,RAC-PK-gamma,RAC-gamma,STK-2
Kit Contents Blocking Buffer, 20X Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, DyLight™ 680 Goat Anti-Mouse IgG (H+L), DyLight 800 Goat Anti-Rabbit IgG (H+L), Thin Plate Seal Assembly, Components included only in the target-specific kits:Antibody #1 and Antibody #2
Label or Dye DyLight™ 680, DyLight™ 800
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Supernatant, 10,000 cells
Storage Requirements 2°C to 8°C
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 24 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.